siRNA Details
| Gene Symbol: DKFZp686K1684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_033971.1 | 3 | 787 | |||
| GenBank mRNA | BX648962.1 | - | LIT3414 Pax6OS |
3 | 787 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

DKFZp686K1684| Gene Symbol: DKFZp686K1684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_033971.1 | 3 | 787 | |||
| GenBank mRNA | BX648962.1 | - | LIT3414 Pax6OS |
3 | 787 | |