siRNA Details
Gene Symbol: LINC00167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024233.1 | 1 | 1286 | |||
GenBank mRNA | AK055956.1 | - | 1 | 1286 | ||
BC137306.1 | - | 1 | 972 | |||
BC137307.1 | - | 1 | 972 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024233.1 | 1 | 1286 | |||
GenBank mRNA | AK055956.1 | - | 1 | 1286 | ||
BC137306.1 | - | 1 | 972 | |||
BC137307.1 | - | 1 | 972 |