siRNA Details
| Gene Symbol: CRACR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144958.1 | NP_001138430.1 | 2 | 249 | ||
| NM_032680.3 | NP_116069.1 | 2 | 249 | |||
| XM_006719021.3 | XP_006719084.1 | 2 | 249 | |||
| GenBank mRNA | AK057608.1 | - | 2 | 249 | ||
| AK304017.1 | - | 2 | 151 | |||
| BC004524.2 | - | 2 | 146 | |||
| CR627161.1 | - | 2 | 318 | |||
| DB214079.1 | - | 2 | 249 | |||

CRACR2A
Genomic Map