siRNA Details
| Gene Symbol: SMCO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013698.2 | NP_001013720.2 | 2 | 127 | ||
| XM_017019312.1 | XP_016874801.1 | 3 | 2792 | |||
| GenBank mRNA | AK092637.1 | - | 2 | 127 | ||
| BC132752.1 | - | 2 | 103 | 40146774 | ||
| BC132754.1 | - | 2 | 103 | 40146776 | ||

SMCO3
Genomic Map