siRNA Details
Gene Symbol: LINC01489 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120466.1 | 2 | 118 | |||
GenBank mRNA | AA056540.1 | - | 3 | 485 | ||
AA056644.1 | - | 2 | 118 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01489 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120466.1 | 2 | 118 | |||
GenBank mRNA | AA056540.1 | - | 3 | 485 | ||
AA056644.1 | - | 2 | 118 |