siRNA Details
| Gene Symbol: LINC01489 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120466.1 | 3 | 259 | |||
| GenBank mRNA | AA056540.1 | - | 2 | 343 | ||
| AA056644.1 | - | 3 | 259 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01489| Gene Symbol: LINC01489 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120466.1 | 3 | 259 | |||
| GenBank mRNA | AA056540.1 | - | 2 | 343 | ||
| AA056644.1 | - | 3 | 259 | |||