siRNA Details
Gene Symbol: LINC00544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033889.1 | 5 | 539 | |||
GenBank mRNA | AI026706.1 | - | 1 | 167 | ||
BC132810.1 | - | 3 | 270 | |||
BC132836.1 | - | 3 | 270 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033889.1 | 5 | 539 | |||
GenBank mRNA | AI026706.1 | - | 1 | 167 | ||
BC132810.1 | - | 3 | 270 | |||
BC132836.1 | - | 3 | 270 |