siRNA Details
| Gene Symbol: LINC00364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130792.1 | 3 | 259 | |||
| GenBank mRNA | BG210290.1 | - | 3 | 259 | ||
| BG216668.1 | - | 2 | 230 | |||
| BG221194.1 | - | 2 | 230 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00364| Gene Symbol: LINC00364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130792.1 | 3 | 259 | |||
| GenBank mRNA | BG210290.1 | - | 3 | 259 | ||
| BG216668.1 | - | 2 | 230 | |||
| BG221194.1 | - | 2 | 230 | |||