siRNA Details
| Gene Symbol: MZT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001071775.2 | NP_001065243.1 | 3 | 1715 | ||
| GenBank mRNA | AK025522.1 | - | 3 | 1588 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MZT1| Gene Symbol: MZT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001071775.2 | NP_001065243.1 | 3 | 1715 | ||
| GenBank mRNA | AK025522.1 | - | 3 | 1588 | ||