siRNA Details
| Gene Symbol: RNASE13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012264.3 | NP_001012264.1 | 2 | 194 | ||
| GenBank mRNA | AY665808.1 | - | 1 | 56 | ||
| BC044831.1 | - | 2 | 194 | 5166630 | ||
| FJ237363.1 | - | 2 | 194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNASE13| Gene Symbol: RNASE13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012264.3 | NP_001012264.1 | 2 | 194 | ||
| GenBank mRNA | AY665808.1 | - | 1 | 56 | ||
| BC044831.1 | - | 2 | 194 | 5166630 | ||
| FJ237363.1 | - | 2 | 194 | |||