siRNA Details
| Gene Symbol: NUMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BG654811.1 | - | 1 | 31 | ||
| Gene Symbol: LOC105370561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055876.1 | - | 1 | 104 | ||

NUMB
Genomic Map