siRNA Details
| Gene Symbol: ZDHHC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006720119.3 | XP_006720182.1 | 1 | 267 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZDHHC22| Gene Symbol: ZDHHC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006720119.3 | XP_006720182.1 | 1 | 267 | ||