siRNA Details
| Gene Symbol: CCDC88C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080414.3 | NP_001073883.2 | 22 | 4015 | ||
| XM_005267691.4 | XP_005267748.1 | 22 | 4669 | |||
| XM_011536796.2 | XP_011535098.1 | 22 | 3969 | |||
| XM_017021335.1 | XP_016876824.1 | 22 | 4669 | |||
| XM_017021336.1 | XP_016876825.1 | 7 | 1078 | |||

CCDC88C
Genomic Map