siRNA Details
| Gene Symbol: MIR381HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104192.1 | 2 | 113 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR381HG| Gene Symbol: MIR381HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104192.1 | 2 | 113 | |||