siRNA Details
| Gene Symbol: GOLGA8J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK302171.1 | - | 1 | 571 | ||
| Gene Symbol: GOLGA8M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005268284.4 | XP_005268341.1 | 1 | 570 | ||
| XM_005268285.4 | XP_005268342.1 | 1 | 570 | |||
| XM_011521952.2 | XP_011520254.1 | 1 | 570 | |||
| XM_011521953.2 | XP_011520255.1 | 1 | 570 | |||

GOLGA8J
Genomic Map