siRNA Details
Gene Symbol: CSNK1A1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027320.1 | 1 | 254 | |||
GenBank mRNA | BC028192.1 | - | 1 | 207 | ||
BC066972.1 | - | 1 | 254 | |||
BG723784.1 | - | 1 | 263 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CSNK1A1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027320.1 | 1 | 254 | |||
GenBank mRNA | BC028192.1 | - | 1 | 207 | ||
BC066972.1 | - | 1 | 254 | |||
BG723784.1 | - | 1 | 263 |