siRNA Details
| Gene Symbol: UNC13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080534.1 | NP_001074003.1 | 1 | 2683 | ||
| XM_005254394.4 | XP_005254451.1 | 2 | 3634 | |||
| XM_017022220.1 | XP_016877709.1 | 6 | 3554 | |||
| XM_017022221.1 | XP_016877710.1 | 5 | 3434 | |||
| XM_017022222.1 | XP_016877711.1 | 4 | 3332 | |||
| XM_017022223.1 | XP_016877712.1 | 3 | 8866 | |||
| XM_017022224.1 | XP_016877713.1 | 2 | 3566 | |||
| XM_017022225.1 | XP_016877714.1 | 1 | 1073 | |||

UNC13C
Genomic Map