siRNA Details
| Gene Symbol: TTLL13P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104604.1 | 6 | 884 | |||
| GenBank mRNA | AK127965.1 | - | 6 | 860 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTLL13P| Gene Symbol: TTLL13P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104604.1 | 6 | 884 | |||
| GenBank mRNA | AK127965.1 | - | 6 | 860 | ||