siRNA Details
Gene Symbol: LINC01578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037601.1 | 1, 2 | 351 | |||
GenBank mRNA | AA083786.1 | - | 1, 2 | 31 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037601.1 | 1, 2 | 351 | |||
GenBank mRNA | AA083786.1 | - | 1, 2 | 31 |