siRNA Details
| Gene Symbol: SHISA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145205.1 | NP_001138677.2 | 2 | 1091 | ||
| GenBank mRNA | AK309038.1 | - | 2 | 432 | ||
| CR627300.1 | - | 2 | 287 | |||
| CR749545.1 | - | 2 | 287 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SHISA9| Gene Symbol: SHISA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145205.1 | NP_001138677.2 | 2 | 1091 | ||
| GenBank mRNA | AK309038.1 | - | 2 | 432 | ||
| CR627300.1 | - | 2 | 287 | |||
| CR749545.1 | - | 2 | 287 | |||