siRNA Details
| Gene Symbol: FRG2KP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135055.1 | 3 | 477 | |||
| GenBank mRNA | BI521010.1 | - | 511 | |||
| Gene Symbol: LOC107983990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005255752.3 | XP_005255809.1 | 3 | 562 | ||

FRG2KP
Genomic Map