siRNA Details
| Gene Symbol: LINC01568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042734.1 | - | 2 | 583 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01568| Gene Symbol: LINC01568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042734.1 | - | 2 | 583 | ||