siRNA Details
| Gene Symbol: RNASEK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004333.4 | NP_001004333.2 | 1, 2 | 348 | ||
| GenBank mRNA | AA633512.1 | - | 2, 3 | 457 | ||
| AA843796.1 | - | 2, 3 | 466 | |||
| AK289930.1 | - | 1, 2 | 131 | |||
| AM746459.1 | - | 1, 2 | 115 | |||
| BC051802.1 | - | 1, 2 | 119 | 4346419 | ||
| BC062705.1 | - | 1, 2 | 123 | 4273688 | ||
| BM835723.1 | - | 1, 2 | 348 | |||
| CB160051.1 | - | 1, 2 | 286 | |||

RNASEK
Genomic Map