siRNA Details
| Gene Symbol: UBE2S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014501.2 | NP_055316.2 | 4 | 1096 | ||
| GenBank mRNA | AB062397.1 | - | 1 | 906 | ||
| BC004236.2 | - | 1 | 906 | 2988449 | ||
| BC007554.1 | - | 1 | 906 | 2967547 | ||
| BC065364.1 | - | 1 | 916 | 5434942 | ||
| BC066948.1 | - | 1 | 752 | 4836851 | ||
| CA421746.1 | - | 1 | 108 | |||
| M91670.1 | - | 4 | 796 | |||

UBE2S
Genomic Map