siRNA Details
| Gene Symbol: LOC79999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291904.1 | NP_001278833.1 | 2, 3 | 252 | ||
| XM_017025119.1 | XP_016880608.1 | 3, 4 | 428 | |||
| GenBank mRNA | BU595835.1 | - | 2, 3 | 263 | ||
| Gene Symbol: LOC102724624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011524126.2 | XP_011522428.1 | 3, 4 | 441 | ||
| Gene Symbol: LOC388436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291905.1 | NP_001278834.1 | 2, 3 | 252 | ||
| GenBank mRNA | BC058012.1 | - | 2, 3 | 264 | ||

LOC79999
Genomic Map