siRNA Details
| Gene Symbol: TBC1D3P5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033892.1 | 12 | 1996 | |||
| GenBank mRNA | BC112354.1 | - | 7 | 2455 | 40034438 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBC1D3P5| Gene Symbol: TBC1D3P5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033892.1 | 12 | 1996 | |||
| GenBank mRNA | BC112354.1 | - | 7 | 2455 | 40034438 | |