siRNA Details
| Gene Symbol: TBC1D3P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027486.1 | 10 | 786 | |||
| GenBank mRNA | AK301746.1 | - | 10 | 787 | ||
| BC058890.1 | - | 10 | 811 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBC1D3P2| Gene Symbol: TBC1D3P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027486.1 | 10 | 786 | |||
| GenBank mRNA | AK301746.1 | - | 10 | 787 | ||
| BC058890.1 | - | 10 | 811 | |||