siRNA Details
| Gene Symbol: TLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011524217.2 | XP_011522519.1 | 1 | 54 | ||
| XM_011524225.2 | XP_011522527.1 | 1 | 54 | |||
| XM_011524228.2 | XP_011522530.1 | 1 | 39 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TLK2| Gene Symbol: TLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011524217.2 | XP_011522519.1 | 1 | 54 | ||
| XM_011524225.2 | XP_011522527.1 | 1 | 54 | |||
| XM_011524228.2 | XP_011522530.1 | 1 | 39 | |||