siRNA Details
| Gene Symbol: LINC00674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027418.1 | 6 | 10020 | |||
| GenBank mRNA | AK091150.1 | - | 1 | 1557 | ||
| AK092798.1 | - | 1 | 741 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00674| Gene Symbol: LINC00674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027418.1 | 6 | 10020 | |||
| GenBank mRNA | AK091150.1 | - | 1 | 1557 | ||
| AK092798.1 | - | 1 | 741 | |||