siRNA Details
| Gene Symbol: MYL12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303047.1 | NP_001289976.1 | 1 | 190 | ||
| NM_006471.3 | NP_006462.1 | 1 | 190 | |||
| GenBank mRNA | BC031972.1 | - | 1 | 190 | 4836883 | |
| EU794621.1 | - | 1 | 190 | |||

MYL12A
Genomic Map