siRNA Details
| Gene Symbol: C19orf71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135580.1 | NP_001129052.1 | 2 | 300 | ||
| XM_011527596.2 | XP_011525898.1 | 2 | 226 | |||
| GenBank mRNA | AI138384.1 | - | 3 | 370 | ||
| BX396051.2 | - | 2 | 304 | |||
| DY654838.1 | - | 3 | 219 | |||

C19orf71
Genomic Map