siRNA Details
| Gene Symbol: LOC440570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135765.1 | 1 | 1038 | |||
| GenBank mRNA | AK125737.1 | - | 1 | 1039 | ||
| BC126340.1 | - | 1 | 223 | 8992056 | ||
| Gene Symbol: FAM231B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_016999988.1 | XP_016855477.1 | 1 | 129 | ||

LOC440570
Genomic Map