siRNA Details
| Gene Symbol: MINOS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001032363.3 | NP_001027535.1 | 1, 2 | 124 | ||
| NM_001204083.1 | NP_001191012.1 | 1, 2 | 124 | |||
| GenBank mRNA | AK094318.1 | - | 1, 2 | 101 | ||
| BC070388.1 | - | 1, 2 | 111 | 30407082 | ||
| BG687187.1 | - | 132 | ||||
| CB157536.1 | - | 1, 2 | 60 | |||
| DA523181.1 | - | 1, 2 | 124 | |||

MINOS1
Genomic Map