siRNA Details
| Gene Symbol: SLC2A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056786.1 | - | 3 | 2171 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC2A1-AS1| Gene Symbol: SLC2A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056786.1 | - | 3 | 2171 | ||