siRNA Details
| Gene Symbol: SLC35A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005270691.4 | XP_005270748.1 | 1 | 205 | ||
| XM_017000869.1 | XP_016856358.1 | 1 | 205 | |||
| XM_017000870.1 | XP_016856359.1 | 1 | 205 | |||
| XM_017000871.1 | XP_016856360.1 | 1 | 205 | |||
| XM_017000872.1 | XP_016856361.1 | 1 | 205 | |||
| Gene Symbol: LOC730081 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097502.1 | - | 1 | 400 | ||

SLC35A3
Genomic Map