siRNA Details
| Gene Symbol: SLC6A17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010898.2 | NP_001010898.1 | 2 | 697 | ||
| XM_006710643.2 | XP_006710706.1 | 2 | 405 | |||
| GenBank mRNA | AK289982.1 | - | 2 | 693 | ||
| BC140908.1 | - | 1 | 282 | 9056720 | ||
| DA081361.1 | - | 2 | 366 | |||
| DA325382.1 | - | 2 | 405 | |||

SLC6A17
Genomic Map