siRNA Details
| Gene Symbol: LOC100996263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121626.1 | 2 | 186 | |||
| GenBank mRNA | BU568889.1 | - | 2 | 278 | ||
| BX088863.1 | - | 2 | 127 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996263| Gene Symbol: LOC100996263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121626.1 | 2 | 186 | |||
| GenBank mRNA | BU568889.1 | - | 2 | 278 | ||
| BX088863.1 | - | 2 | 127 | |||