siRNA Details
| Gene Symbol: RGS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045630.1 | 4 | 1315 | |||
| GenBank mRNA | AA805263.1 | - | 1 | 192 | ||
| BC032239.1 | - | 4 | 1276 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGS5| Gene Symbol: RGS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045630.1 | 4 | 1315 | |||
| GenBank mRNA | AA805263.1 | - | 1 | 192 | ||
| BC032239.1 | - | 4 | 1276 | |||