siRNA Details
| Gene Symbol: CNIH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322302.1 | NP_001309231.1 | 1 | 238 | ||
| NM_001322303.1 | NP_001309232.1 | 1 | 238 | |||
| NM_001322304.1 | NP_001309233.1 | 1 | 238 | |||
| NR_136291.1 | 1 | 238 | ||||
| NR_136292.1 | 1 | 238 | ||||
| NR_136293.1 | 1 | 238 | ||||
| NR_136294.1 | 1 | 238 | ||||
| NR_136295.1 | 1 | 238 | ||||
| NR_136296.1 | 1 | 238 | ||||
| NR_136297.1 | 1 | 238 | ||||

CNIH3
Genomic Map