siRNA Details
| Gene Symbol: PCNX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014801.3 | NP_055616.3 | 1 | 318 | ||
| XM_006711816.3 | XP_006711879.1 | 1 | 140 | |||
| GenBank mRNA | CT004972.1 | - | 1 | 318 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCNX2| Gene Symbol: PCNX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014801.3 | NP_055616.3 | 1 | 318 | ||
| XM_006711816.3 | XP_006711879.1 | 1 | 140 | |||
| GenBank mRNA | CT004972.1 | - | 1 | 318 | ||