siRNA Details
| Gene Symbol: LOC148413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015434.1 | 1 | 101 | |||
| GenBank mRNA | AK024040.1 | - | 1 | 79 | ||
| AK297832.1 | - | 1 | 90 | |||
| BC068600.1 | - | 1 | 62 | |||
| BC080546.1 | - | 1 | 57 | |||
| BC104242.1 | - | 1 | 105 | |||
| BC104243.1 | - | 1 | 101 | |||
| BC112998.1 | - | 1 | 99 | |||
| BC112999.1 | - | 1 | 99 | |||

LOC148413
Genomic Map