siRNA Details
Gene Symbol: APCDD1L-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034147.1 | 7 | 1198 | |||
GenBank mRNA | AK054637.1 | - | 7 | 1200 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: APCDD1L-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034147.1 | 7 | 1198 | |||
GenBank mRNA | AK054637.1 | - | 7 | 1200 |