siRNA Details
| Gene Symbol: ANKRD20A8P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003366.2 | 20, 21 | 3391 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD20A8P| Gene Symbol: ANKRD20A8P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003366.2 | 20, 21 | 3391 | |||