siRNA Details
| Gene Symbol: DSCR4-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AW664531.1 | - | 1 | 229 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DSCR4-IT1| Gene Symbol: DSCR4-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AW664531.1 | - | 1 | 229 | ||