siRNA Details
Gene Symbol: DGCR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024159.1 | 1 | 1923 | |||
GenBank mRNA | BC036821.1 | - | 1 | 1934 | ||
L77571.1 | - | 1 | 1821 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DGCR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024159.1 | 1 | 1923 | |||
GenBank mRNA | BC036821.1 | - | 1 | 1934 | ||
L77571.1 | - | 1 | 1821 |