siRNA Details
| Gene Symbol: LINC00895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024381.1 | 1 | 413 | |||
| GenBank mRNA | AK057431.1 | - | 1 | 413 | ||
| BC042984.2 | - | 1 | 381 | |||
| BC104250.1 | - | 1 | 355 | |||
| BC104251.1 | - | 1 | 352 | |||
| BC113002.1 | - | 1 | 355 | |||
| BC113003.1 | - | 1 | 355 | |||

LINC00895
Genomic Map