siRNA Details
Gene Symbol: LOC107985555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029168.1 | XP_016884657.1 | 4 | 634 | ||
XM_017029169.1 | XP_016884658.1 | 3 | 763 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC107985555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029168.1 | XP_016884657.1 | 4 | 634 | ||
XM_017029169.1 | XP_016884658.1 | 3 | 763 |