siRNA Details
| Gene Symbol: LOC105374013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011513341.2 | XP_011511643.1 | 2 | 2191 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105374013| Gene Symbol: LOC105374013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011513341.2 | XP_011511643.1 | 2 | 2191 | ||