siRNA Details
| Gene Symbol: HS1BP3-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046836.1 | 2 | 287 | |||
| GenBank mRNA | BQ010717.1 | - | 1 | 326 | ||
| BQ013798.1 | - | 1 | 326 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HS1BP3-IT1| Gene Symbol: HS1BP3-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046836.1 | 2 | 287 | |||
| GenBank mRNA | BQ010717.1 | - | 1 | 326 | ||
| BQ013798.1 | - | 1 | 326 | |||