siRNA Details
| Gene Symbol: LOC100499194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034130.1 | 1 | 478 | |||
| GenBank mRNA | BG772716.1 | - | 2 | 286 | ||
| DB024523.1 | - | 509 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100499194| Gene Symbol: LOC100499194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034130.1 | 1 | 478 | |||
| GenBank mRNA | BG772716.1 | - | 2 | 286 | ||
| DB024523.1 | - | 509 | ||||